DuPont™ Tychem™ 10,000 Level A Suits
Supplier:  DuPont™ TK554TLYXL000100
 View more versions of this product
Catalog No. 29-636-1468
Explore available promotions
Description
Description
Fully Encapsulated Level A Suits are ideal for industrial, HazMat and domestic preparedness situations. High-visibility lime yellow.
  • Double taped seams
  • Three-layer faceshield (PVC 40mil/PTFE 5mil/PVC 20mil)
  • Gas-tight PVC zipper closure
  • Double storm flaps with Velcro™ closure
  • Attached internal Barrier gloves, outer butyl gloves and sock boots with outer boot flaps
  • Knee wear pads
  • Two exhaust valves
  • Internal adjustment belt
  • Garments successfully tested against more than 260 challenge chemicals
Compliance
Meets OSHA 1910.132 (a) standards.
Specifications
Specifications
Lime Yellow
Tychem
Gas-tight zipper and double storm flaps with hook-and-loop closure
5.9 to 6.3 ft.
30.75 in.
78.105 cm
Butyl gloves
46.75 to 50.25 in.
Front entry
Industrial, HazMat and domestic preparedness applications
X-Large
Expanded faceshield, 2 exhaust valves, internal adjustment waist belt, attached internal multilayer laminate/external Butyl size 10 gloves, socks with outer boot flaps, soles; Specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals, excellent chemical barrier properties and offers an extremely durable Fabric that is puncture and tear-resistant
1.79 to 1.92 m
29.25 in.
74.29 cm
Taped
118.74 to 127.65 cm
27 mil