DuPont™ Tychem™ 2000 Series 125 Coveralls
Flexible and durable light liquid splash protection
Supplier:  DuPont™ QC125SYL5X001200
 View more versions of this product
Catalog No. 191202582
Explore available promotions
Description
Description
Tychem™ QC is polyethylene-coated Tyvek™ fabric fabric provides at least 30 minutes of protection against 42 chemical challenges
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Yellow for high visibility
  • 10 mil thickness
QC 125B
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
QC 125S
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • QC 125T
    • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
    • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
    Compliance
    Meets ASTM™ D3776, ASTM D1777, ASTM 1117, ASTM D3786, ASTM D5034 standards.
    Specifications
    Specifications
    5X-Large
    Yellow
    Elastic
    Zipper
    Tyvek™ Fabric with Polyethylene Coated
    33 in.
    Petroleum refining, pulp and Paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
    102.87 cm
    6.7 to 6.10 ft.
    Tychem
    Elastic
    Collared
    Serged
    53.75 to 57.25 in.
    136.52 to 145.41 cm
    40.5 in.
    33.5 in.
    2.04 to 1.85 m