DuPont™ Tychem™ 2000 Series 125 Coveralls
Flexible and durable light liquid splash protection
Supplier:  DuPont™ QC125TYL2X000400
 View more versions of this product
Catalog No. 19-162-756
Explore available promotions
Description
Description
Tychem™ QC is polyethylene-coated Tyvek™ fabric fabric provides at least 30 minutes of protection against 42 chemical challenges
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Yellow for high visibility
  • 10 mil thickness
QC 125B
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
QC 125S
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • QC 125T
    • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
    • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
    Compliance
    Meets ASTM™ D3776, ASTM D1777, ASTM 1117, ASTM D3786, ASTM D5034 standards.
    Specifications
    Specifications
    2X-Large
    Yellow
    Front Zipper
    Petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing
    Tychem
    Collared
    Taped