DuPont™ Tychem™ 2000 Series 127 Coveralls
Flexible and durable light liquid splash protection
Supplier:  DuPont™ QC127TYL2X000400
 View more versions of this product
Catalog No. 19-088-1244B
Explore available promotions
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments. Also garments made of Tyvek should have slip-resistant or antislip materials on the outer surface of boots, shoe covers, or other garments in conditions where slipping could occur.
Description
Description
Tychem™ QC is polyethylene-coated Tyvek™ fabric fabric provides at least 30 minutes of protection against 42 chemical challenges
  • Attached hood with elastic around face opening
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Yellow for high visibility
  • 10 mil thickness
QC 127B
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
QC 127S
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • QC 127T
    • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
    • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
    Compliance
    Meets ASTM™ D3776, ASTM D3786, ASTM D1117, ASTM D5034 standards.
    Specifications
    Specifications
    2X-Large
    Yellow
    Elastic
    Standard
    Taped
    30 in.
    Petroleum refining, pulp and Paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
    97.15 cm
    6 to 6.4 ft.
    Tychem
    Elastic
    Hooded
    Zipper
    Polyethylene Film Coated on Tyvek™ Fabric
    112.39 to 121.28 cm
    38.25 in.
    28.75 in.
    1.82 to 1.95 m