DuPont™ Tychem™ 2000 SFR Apron
Tychem™ 2000 SFR Aprons provide chemical and secondary flame protection in a lightweight garment. Tychem™ 2000 SFR Aprons are intended to be worn over primary flame resistant garments.
Supplier:  DuPont™ QS275TGR2X001200
 View more versions of this product
Catalog No. 17-986-626
Explore available promotions
Description
Description
  • Tychem™ 2000 SFR represents a new generation of secondary flame-resistant chemical garment technology
  • Unlike traditional secondary flame resistant chemical garments that have been available for years, new Tychem™ 2000 SFR garments were specially designed to meet dual hazard needs of a protective chemical suit with secondary flame resistance
  • This unique combination of performance permits Tychem™ 2000 SFR garments to be worn over primary flame resistant (FR) garments like Nomex™ when chemical splash and flash fire hazards exist
  • The fabric used in Tychem™ 2000 SFR garments utilize a unique technology so that it does not char like traditional secondary flame resistant technologies - it is designed to shrink away from flame without burning
  • Tychem™ 2000 SFR garments also provide an effective barrier against a range of inorganic acids, bases, industrial cleaning chemicals, and particles
  • They're an ideal option for keeping workers safe from chemicals, while not compromising the flame protection of fire-resistance clothing worn beneath them
  • Applications include: refineries, petrochemical plants, laboratories, hazardous maintenance operations
Specifications
Specifications
2X-Large
Single-use
Non-sterile
Yes
45.75 in.
112 cm
Green
Hook and Loop, Waist Ties
Elastic
7.5 mil
33 in.
Taped