missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tychem™ 4000 Series 127 Coveralls
Ideal for chemical mixing, remediation, emergency medical response, paint spraying, and radioactive environments
Supplier: DuPont™ SL127TWHLG000600
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Lightweight, comfortable garment designed for easy wear. Excellent for waste management, PCB cleanup, hazardous response, and nuclear environments.
- DuPont™Tyvek™ fabric laminated with Dow™ Saranex™-23P film
- Attached hood
- Elasticized face, wrists, and ankles
- Zipper front with storm flap
- Bound-seam models are tightly sewn with a reinforced outer binding to increase strength and particle barrier
- Taped-seam models are sewn and taped to provide chemical resistance against heavy liquid splashes and tough seam stress
Compliance
Meets ASTM™ D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786; 16 CFR 1610 Standard for Flammability of Clothing Textiles; MIL-STD-282 requirements.
Specifications
Large | |
White | |
Elastic | |
Front Zipper | |
ASTM D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786, 16 CFR 1610, MIL-STD-282 | |
97.15 to 106.04 cm | |
Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments | |
88.9 cm | |
5.5 to 5.9 ft. | |
0.22 lb. |
Tychem | |
Elastic | |
Hooded | |
Taped | |
29 in. | |
12 mil | |
35 in. | |
25.75 in. | |
1.67 to 1.8 m | |
0.1 kg |