DuPont™ Tychem™ 5000 Encapsulated Level B Suit, Expanded Back
DuPont™ Tychem™ 5000 Encapsulated Level B Suit, Expanded Back provides protection against a broad range of chemicals and situations involving liquid splash.
Supplier: DuPont™ C3528TTNMD000600
Description
DuPont™ Tychem™ 5000 helps offer protection for those in hazardous environments.
- Provides at least 30 minutes of protection against over 150 chemical challenges
- NFPA 1992 certified