Learn More
DuPont™ Tychem™ 6000FR Coverall with Elastic Wrist, Draw String Hood, Sock Boots, Boot Flaps
Tychem™ 6000 FR offers triple-hazard protection from liquid-chemical splash, flash fire, and electric arc.
Supplier: DuPont™ TP199TOR5X000200
Description
Tychem™ 6000 FR coveralls combine the trusted chemical protection of Tychem™ fabric and flame protection of Nomex™ fiber in a single-layer garment. Tychem™ 6000 FR garments meet the requirements of NFPA 1992 Standard on Liquid-Splash Protective Ensembles and Clothing for Hazardous Materials Emergencies and NFPA 2112, Standard on Flame-Resistant Garments for Protection of Industrial Personnel against Flash Fire. Tychem™ 6000 FR also exceeds the Hazard Risk Category 2 requirement of 8 cal/cm2 outlined in NFPA 70E, Standard for Electrical Safety in the Workplace. Tychem™ 6000 FR fabrics are tested by a third party laboratory to assess its chemical permeation resistance.
- Provides at least 30 minutes of protection against over 180 chemical challenges plus the added benefit of electric arc, and flash fire burn injury protection
- Utilized and relied on by industrial workers and HAZMAT responders Available in orange for high visibility and gray for low visibility
Specifications
5X-Large | |
Tychem | |
Single-use | |
Elastic | |
Taped | |
Chemicals, Liquid | |
None |
Unisex | |
Orange | |
Attached Socks, Boot Overflaps | |
Hooded | |
Non-sterile | |
Yes | |
2/Pk. |