DuPont™ Tychem™ 4000 Series 122 Coveralls
Delivers effective protection against a range of chemical environments.
Supplier:  DuPont™ SL122TWH4X000600
 View more versions of this product
Catalog No. 19088348G
Explore available promotions
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
Tychem SL, utilizes Saranex™ 23-P film laminated to Tyvek fabric to provide a rugged and durable fabric that resists abrasion.
The fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Attached hood with elastic around face opening
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
Compliance
Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.
Specifications
Specifications
Hooded Coveralls with Socks
Tyvek with Saranex 23-P Film Laminate
Single-use
Elastic
Attached, Elastic Face
Taped
Attached hood, elastic wrist, attached socks
32 in.
12 mil
39.5 in.
32 in.
6.33 to 6.58 ft.
4X-Large
White
Attached Socks
Hooded
Front Zipper
Non-sterile
50.75 to 54.25 in.
81.28 cm
Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
100.33 cm
81.28 cm
1.92 to 2.01 m