Learn More
Tyramine hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1222006
Description
Reagent for fluorescence enzyme immunoassay
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tyramine hydrochloride | |
271°C to 274°C | |
MFCD00012901 | |
3627058 | |
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147 | |
[H+].[Cl-].NCCC1=CC=C(O)C=C1 | |
173.64 | |
173.65 |
60-19-5 | |
C8H12ClNO | |
5 g | |
14,9835 | |
RNISDHSYKZAWOK-UHFFFAOYSA-N | |
4-(2-aminoethyl)phenol;hydrochloride | |
66449 | |
98% |
Chemical Identifiers
60-19-5 | |
173.64 | |
RNISDHSYKZAWOK-UHFFFAOYSA-N | |
66449 | |
[H+].[Cl-].NCCC1=CC=C(O)C=C1 |
C8H12ClNO | |
MFCD00012901 | |
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147 | |
4-(2-aminoethyl)phenol;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-462-5
RTECSNumber : SJ6050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only