missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tyramine hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1222014

 View more versions of this product

Catalog No. AAA1222014


Explore available promotions

Description

Description

Reagent for fluorescence enzyme immunoassay

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tyramine hydrochloride
271°C to 274°C
MFCD00012901
3627058
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
[H+].[Cl-].NCCC1=CC=C(O)C=C1
173.64
173.65
60-19-5
C8H12ClNO
25 g
14,9835
RNISDHSYKZAWOK-UHFFFAOYSA-N
4-(2-aminoethyl)phenol;hydrochloride
66449
98%

Chemical Identifiers

60-19-5
173.64
RNISDHSYKZAWOK-UHFFFAOYSA-N
66449
[H+].[Cl-].NCCC1=CC=C(O)C=C1
C8H12ClNO
MFCD00012901
tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
4-(2-aminoethyl)phenol;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-462-5

RTECSNumber : SJ6050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only