DuPont™ Tyvek™ 400 Coveralls with Elastic Wrists and Ankles, Vending Packaged
Individually packaged coveralls for PPE vending feature elastic wrists and ankles to provide an ideal balance of protection, durability, and comfort.
Supplier:  DuPont™ TY125SWHLG0025VP
 View more versions of this product
Catalog No. 19-820-525
Explore available promotions
Description
Description
  • Vending packaged, serged seams
  • Tyvek composed of flash spun high density polyethylene which creates a unique, nonwoven material that provides durability and comfort
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Laydown collar; longer zipper extends to chin for complete coverage of neck area
  • Storm flap of garment material covers zipper
  • Elastic opening for tighter fit at wrists and ankles
  • Elastic waist to better position the garment
  • Thumb loop to keep garment sleeve in place
  • Applications include fiberglass and composite manufacturing, wind turbines, boat construction and repair, remediation, and maintenance
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Elastic
Unhooded
Serged
Particle
ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127
Yes
Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning
Individually Packaged for PPE Vending
Large
Tyvek
Limited-use
Elastic
Collared
Front Zipper
Non-sterile
Laydown
Flashspun HDPE
TY125SWH
25/Cs.