DuPont™ Tyvek™ 400 Disposable Pants, USDA, FDA, OSHA
DuPont™ Tyvek™ 400 pants provide the protection and durability you need so you can focus on the task at hand. Meets USDA, FDA, OSHA Standards.
$75.08 - $91.08
Specifications
Color White
Ankle Style Open
Seam Type Serged
Waist Style Elastic
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-500-290
View Documents
DuPont™
TY350SWH2X0012G1
2X-Large
Case of 12 Each for $79.64
17-500-291
View Documents
DuPont™
TY350SWH3X0012G1
3X-Large
Case of 12 Each for $84.31
17-500-292
View Documents
DuPont™
TY350SWH4X0012G1
4X-Large
Case of 12 Each for $91.08
17-500-296
View Documents
DuPont™
TY350SWHLG0012G1
Large
Case of 12 Each for $75.08
17-500-297
View Documents
DuPont™
TY350SWHMD0012G1
Medium
Case of 12 Each for $75.08
17-500-298
View Documents
DuPont™
TY350SWHXL0012G1
X-Large
Case of 12 Each for $75.08
Description
Description
DuPont™ Tyvek™ 400 provides light-weight inherent barrier protection against hazardous dry particles and aerosols, and nonhazardous light liquid splash.
  • Excellent abrasion resistance due its unique, nonwoven material
  • Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
USDA, FDA, OSHA
Specifications
Specifications
White
Open
Serged
Elastic
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'