DuPont™ Tyvek™ 400 Disposable Shirt, USDA, FDA, OSHA
DuPont™ Tyvek™ 400 shirts provide the protection and durability you need so you can focus on the task at hand. Meets USDA, FDA, OSHA Standards.
Supplier:  DuPont™ TY303SWH3X0012G1
 View more versions of this product
Catalog No. 17-500-282
Explore available promotions
Description
Description
DuPont™ Tyvek™ 400 provides light-weight inherent barrier protection against hazardous dry particles and aerosols, and nonhazardous light liquid splash.
  • Excellent abrasion resistance due its unique, nonwoven material
  • Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
USDA, FDA, OSHA
Specifications
Specifications
3X-Large
Collar
Snaps
Extends to hip
White
Serged
Open