DuPont™ Tyvek™ 400 Disposable Shirt, USDA, FDA, OSHA
DuPont™ Tyvek™ 400 shirts provide the protection and durability you need so you can focus on the task at hand. Meets USDA, FDA, OSHA Standards.
Supplier: DuPont™ TY303SWHMD0012G1
Description
DuPont™ Tyvek™ 400 provides light-weight inherent barrier protection against hazardous dry particles and aerosols, and nonhazardous light liquid splash.
- Excellent abrasion resistance due its unique, nonwoven material
- Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
USDA, FDA, OSHA
Specifications
Medium | |
Collar | |
Snaps | |
Extends to hip |
White | |
Serged | |
Open |