DuPont™ Tyvek™ 400 Frocks, Mandarin Collar
Flash-spun high-density polyethylene provides an ideal balance of protection, durability, and comfort
Supplier:  DuPont™ TY216SWH3X003000
 View more versions of this product
Catalog No. 19-067-052
Explore available promotions
Description
Description
Ideal for lead and asbestos abatement and remediation, general maintenance and operations, spray painting, general clean-up
  • Inherent barrier to small size particles built into the fabric itself — no films or laminates to abrade or wear away
  • Body-length jacket with five front snap closures for easy donning and doffing
  • Enhanced frontal protection
  • Mandarin collar fits tightly around hooded PAPR
  • Elastic opening for tighter fit at wrist
  • Serged seams
  • White
Specifications
Specifications
General Purpose Frock
Tyvek
Limited-use
Elastic
Lead and asbestos abatement and remedation, general maintenance and operations, spray painting, general clean-up
48.75 to 52.25 in.
113.03 cm
3X-Large
White
Snaps
Serged
Mandarin
44.5 in.