DuPont™ Tyvek™ 400 Boot Covers
High boot covers with elastic top offers an inherent barrier against particles and provides durability even after wear and abrasion.
Supplier:  DuPont™ TY454SWHXL0100SR
 View more versions of this product
Catalog No. 19-813-2157
Explore available promotions
Alert:
Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
High boot covers with elastic top offers an inherent barrier against particles and provides durability even after wear and abrasion.
  • Abrasion- and tear-resistant
  • Lightweight and comfortable
  • Tyvek 400 fabric offers an inherent barrier against particles down to 1.0 micron in size
  • Serged seams
  • Elastic opening at calf, elastic ankle
  • FC skid-resistant sole
  • Height: 18 in. (46 cm)
Specifications
Specifications
X-Large
White
Non-sterile
Anti-skid
Serged
Lead and Asbestos Abatement, Remediation, General Maintenance, Spray Painting, Cleaning
40.64 cm
45.72 cm
Tyvek
Single-use
Elastic Top and Ankles
TY454SWHXL0100SR
5.9 mil
16 in.
18 in.
Boot Covers