DuPont™ Tyvek™ 500 Hooded Coveralls
Tyvek™ 500 Hooded Anti-static Coveralls boast stormflaps and serged external seams, with additional protection against blood and bodily fluids.
$317.35 - $356.54
Specifications
Material Tyvek
Color White
Waist Style Elastic (Glued-in)
Ankle Style Elastic
Cuff Style Elastic
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-067-050
View Documents
DuPont™
TY198SWHMD0025LA
Medium
Case of 25 Each for $317.35
19-067-049
View Documents
DuPont™
TY198SWHLG0025LA
Large
Case of 25 Each for $317.35
19-067-051
View Documents
DuPont™
TY198SWHXL0025LA
X-Large
Case of 25 Each for $317.35
19-067-047
View Documents
DuPont™
TY198SWH2X0025LA
2X-Large
Case of 25 Each for $336.38
19-067-048
View Documents
DuPont™
TY198SWH3X0025LA
3X-Large
Case of 25 Each for $356.54
Description
Description
  • Chemical protective clothing, Category III, Type 5-B and 6-B.
  • EN 1073-2 (protection against radioactive contamination)
  • Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603)
  • Antistatic treatment (EN 1149-5) on both sides
  • Serged external seams featuring patent-pending technology for enhanced liquid spray tightness
  • Very low inward leakage due to optimized design
  • Tyvek™ auto-lock zipper and storm flap for increased protection
  • Elastic face, wrists and ankles as well as glued-in waist elastic
Specifications
Specifications
Tyvek
Elastic (Glued-in)
Elastic
Storm Flap
TY198SWH
White
Elastic
Elastic
Serged
25 Coveralls/Cs.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'