DuPont™ Tyvek™ 500 HV Coveralls with Hood
Tyvek™ 500 HV garments are mainly composed of flash spun polyethylene, providing an ideal balance of protection, durability and comfort. Thanks to their nonwoven structure, they are permeable to both air and water vapor, yet repel water-based liquids and aerosols.
Supplier: DuPont™ TY127SHV3X0025XC
Description
- EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), EN ISO 20471 (High Visibility Clothing, Class 3)
- Antistatic treatment (EN 1149-5) on inside
- Stitched external seams
- Very low inward leakage thanks to optimized design
- Tyvek™ auto-lock zipper and zipper flap for increased protection
- Tyvek™ 500 HV meets the single-use disposable coverall requirements of ANSI/ISEA 107-2020 Type R/Class 3 Section 11
Specifications
3X-Large | |
Fluorescent Orange | |
Elastic | |
Elastic, Respirator Fit | |
Serged |
Tyvek 500 | |
Elastic | |
Elastic | |
Zipper | |
25 Coveralls/Pk. |