missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Clean Processed Frocks
Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Clean processed garments offer lowest level of particle shedding within DuPont product portfolio.
Supplier: DuPont™ IC264SWHSM00300C
Alert:
Should not be worn around open flame, heat source, sparks or ignition sources.
Description
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
- Elastic openings for tighter fit at wrist
- Bound neck for lower particle shedding
- Front zipper closure for easy donning and doffing
- Raglan sleeve design for greater range of motion and comfort
- A-line silhouette
Specifications
Controlled Environments Frock | |
Flashspun Polyethylene | |
Non-sterile | |
Elastic | |
Bound | |
81.3 to 91.4 cm | |
Individually Packaged | |
34 in. |
Small | |
White | |
Zipper | |
Serged | |
32 to 36 in. | |
Raglan Sleeve | |
41 in. |