missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ Tyvek™ IsoClean™ Clean Processed Frocks

Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Clean processed garments offer lowest level of particle shedding within DuPont product portfolio.

Supplier:  DuPont™ IC264SWHLG00300C

 View more versions of this product

Catalog No. 17-990-522


Explore available promotions

Alert:

Should not be worn around open flame, heat source, sparks or ignition sources.


Description

Description

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Elastic openings for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • A-line silhouette

 

Specifications

Specifications

Controlled Environments Frock
Flashspun Polyethylene
Non-sterile
Elastic
Bound
91.4 to 96.5 cm
Individually Packaged
114.3 cm
95.25 cm
Large
White
Zipper
Serged
36 to 38 in.
Raglan Sleeve
45 in.
37.5 in.
Videos
SDS
Documents

Documents

Promotions

Promotions