Learn More
Ibidi Usa Inc -Slide VI 0.4, ibiTreat - Tissue Culture Treated Coated Polymer Coverslip
Supplier: Ibidi Gmbh 80606
The ibidi product family is comprised of a variety of -Slides and -Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The convenient six channel format of the -Slide VI 0.4 is ideal for static cell cultivation and the application of standard immunofluorescence protocols, like treatment, staining, and microscopy of living or fixed cells. Alternatively, the -Slide VI 0.4 can be connected to a pump and enables you to observe cells under flow conditions. 6 x 400 m high channels, #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box