Learn More
Invitrogen™ UltraComp eBeads™ Compensation Beads
Designed for use in compensation with all fluorochromes excited by ultraviolet, violet, blue, green, yellow-green, and red lasers. UltraComp eBeads™ react with antibodies of mouse, rat, and hamster origin, and are immunoglobulin light chain-independent. 1 drop (50μL)/test.
Supplier:  Invitrogen™ 01222242
 View more versions of this product
Catalog No. 501129040
Only null left
Explore available promotions
Alert:
Contains Sodium azide
Description
Description
  • Each drop of beads contains both a positive population that will capture any mouse, rat or hamster antibody and a negative population that will not react with antibody
  • This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments
  • Compatible with all fluorochromes excited by an ultraviolet (355nm) or violet (405nm) laser
  • Should be used with standard staining buffers which contain PBS or HBSS, protein such as BSA or FBS, and sodium azide. No other additives should be used.
Specifications
Specifications
Flow Cytometer
100 tests
Compensation Beads
100 Tests
eBeads
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.