Invitrogen™ UltraComp eBeads™ Plus Compensation Beads
UltraComp eBeads Plus Compensation Beads are the second generation of UltraComp beads for use in flow cytometry.
Supplier: Invitrogen™ 01333341
Description
- Binds the spherical particles for use as a single-color compensation controls.
- Binds with mouse, rat and hamster antibodies, but also bind with rabbit and recombinant human antibodies.
- Designed for use in compensation with all fluorochromes excited by ultraviolet (355 nm) violet (405 nm), blue (488 nm), green (532 nm), yellow-green (561 nm), and red (633-640 nm) lasers.
- Compensate in a manner similar to cells when used with Brilliant Violet 785/786-, Brilliant Violet 711-, Super Bright 780- and Super Bright 702-conjugated antibodies.
- Multispecies compatibility—bind with mouse, rat, hamster, rabbit and recombinant human antibodies
- Improved fluorophore compatibility
- Each drop of beads contains two populations: a positive population that bind with any mouse, rat, hamster, rabbit or recombinant human antibody and a negative population that will not bind with antibody.
- When a fluorochrome-conjugated antibody is added to the beads, both positive and negative populations are observed.
- This bimodal distribution can be used for single-color compensation controls to accurately setup instrumentation for multicolor flow cytometry experiments.
Specifications
Compensation Beads | |
Flow Cytometry | |
25 tests | |
eBeads™ |
RUO use only.