Fisherbrand™ Ultrasonic Cleaner Perforated Pans
Supports easy removal of items from ultrasonic cleaners.
$145.45 - $145.45
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | For Use With (Equipment) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | For Use With (Equipment) | Price | Quantity | |||
15-335-20A
|
Fisherbrand™
CPN169058 |
94.67 oz. | 2.8 L | Ultrasonic Cleaner, FS203 |
Each for $145.45
|
|
|||
Description
Perforated Pans
- Support items in the tank for direct cleaning
- For easy removal of items from the tank
- Reduce evaporation and eliminate splashing problems
- Beaker positioning covers required to suspend standard 600mL beakers in the tank
- Allows placement of 250 or 400mL beakers within the standard beaker cover
† Discontinued model.
Specifications
Perforated Pan |