missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Biochrom Ultrospec™ 30 Cell Density Meter
Ultrospec™ 30 Cell Density Meter for measuring density of bacteria and yeast cells in suspension.
Supplier: Biochrom 80300801
Description
- Small, portable, and easy to use, the instrument can measure growth rates of E. coli, other bacteria, and yeast
- Create and save cell specific standard curves to read against again and again
- Can also be used with McFarland standards
- Intuitive touch screen for easy plug and play use
- Battery operated and portable for use where cells are actually cultured
- Uses LED light source in combination with an advanced fiber optics
- Hand-held
- Accepts 10 x 10mm cuvettes or 16 mm test tubes. 10 to 12 mm test tubes require adapter set
- Spillages can be easily wiped from the smooth surface, and the cell compartment area can be flushed with formaldehyde or ethylene oxide to sterilize it
- Save results onboard or export via USB flash drive
- Compact, low-maintenance device
- Not recommended for use with mammalian cell lines
Specifications
CE and UKCA Marked, US and CAN Safety (ETL or equivalent) | |
10 x 10 mm Cuvettes, 16 mm Test Tubes | |
CellDensity Meter | |
Photodiode | |
170 x 210 x 50 mm | |
LED | |
Visible Spectrophotometer | |
600 nm | |
Microbiology, Bacteria and Yeast Quantification | |
LED, Diode Array | |
Absorbance 0.3 to 2.0 A | |
USB-C, UK, Euro, USA, China | |
Round |
Black | |
USB-A, USB-C | |
600 nm | |
LCD Touchscreen | |
English, Chinese | |
40 nm | |
5 V | |
600 nm | |
4.3 in., 480 x 272 Pixel Resolution Backlit LCD Color Display | |
< ± 0.05 A, 1 A using Neutral Density Filter | |
± 0.02 A at 1 A om 10 mm Cuvette | |
100 to 240 V, 50/60 Hz, 10 W |
Contains internal rechargeable Li-Ion battery