missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ HDPE, Carboy UN/DOT Approved
For Use With: Storage and secure shipments of liquid waste; Closure Material: HDPE; Color: White
Supplier: Justrite™ JMC12951
Description
- For packaging group II and hazardous chemicals
- HDPE construction for excellent chemical compatibility
Compliance
Meets UN 3H1 standards
Specifications
| 5.3 gal. | |
| UN/DOT | |
| 70 mm | |
| 279 x 235 x 394 mm | |
| 15.5 in. | |
| HDPE | |
| Rectangular |
| 20 L | |
| HDPE | |
| White | |
| Storage and secure shipments of liquid waste | |
| 11 in. | |
| Carboy | |
| 9 in. |