missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UNC84B Recombinant Rabbit Monoclonal Antibody (8E8D7)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA537996
Description
Positive Samples: HeLa, Jurkat Immunogen sequence: RIRPTAVTLE HVPKALSPNS TISSAPKDFA IFGFDEDLQQ EGTLLGKFTY DQDGEPIQTF HFQAPTMATY QVVELRILTN WGHPEYTCIY RFRVHGEPAH.
UNC84B may form a physical interaction between the nuclear envelope and the centrosome.Specifications
| UNC84B | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BJS4, Q9UH99 | |
| SUN2 | |
| A synthetic peptide corresponding to a sequence within amino acids 618-717 of human SUN2 (Q9UH99). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 8E8D7 | |
| Unconjugated | |
| SUN2 | |
| B230369L08Rik; C030011B15; FRIGG; KIAA0668; nuclear envelope protein; protein unc-84 homolog B; Rab5-interacting protein; rab5IP; RGD1563141; Sad1 and UNC84 domain containing 2; Sad1 unc-84 domain protein 2; sad1/unc-84 protein-like 2; SUN domain-containing protein 2; SUN2; unc-84 homolog B; UNC84B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 223697, 25777, 315135 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |