missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Undecafluorohexanoic Acid (ca. 5mmol) 98.0+%, TCI America™

Supplier:  TCI America A57151SAMPLE

Catalog No. A5715SET


Explore available promotions

Description

Description

Specifications

Specifications

Undecafluorohexanoic Acid (ca. 5mmol) [Ion-Pair Reagent for LC-MS]
14°C
157°C
MFCD00198040
3265
PXUULQAPEKKVAH-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoic acid
67542
314.05
Liquid
307-24-4
Colorless
C6HF11O2
Set
IPC-PFFA-6, Perfluorohexanoic Acid
C(=O)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)O
314.054
CHEBI:83492
≥98.0% (T)

Chemical Identifiers

307-24-4
314.054
PXUULQAPEKKVAH-UHFFFAOYSA-N
67542
2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoic acid
C6HF11O2
MFCD00198040
IPC-PFFA-6, Perfluorohexanoic Acid
CHEBI:83492
C(=O)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1182

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions