missing translation for 'onlineSavingsMsg'
Learn More

Undecanoic acid, 99%

CAS: 112-37-8 | C11H22O2 | 186.29 g/mol

$47.72 - $47.72

Chemical Identifiers

CAS 112-37-8
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.29
MDL Number MFCD00002730
InChI Key ZDPHROOEEOARMN-UHFFFAOYSA-N
Synonym hendecanoic acid, undecylic acid, n-undecanoic acid, n-undecoic acid, n-undecylic acid, undecoic acid, undecanoate, 1-decanecarboxylic acid, undekansaeure, unii-138on3iiqg
PubChem CID 8180
ChEBI CHEBI:32368
IUPAC Name undecanoic acid
SMILES CCCCCCCCCCC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC173970050
SDS View Documents
Thermo Scientific Chemicals
173970050
5 g Glass bottle
Each for $47.72
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-37-8
186.29
ZDPHROOEEOARMN-UHFFFAOYSA-N
8180
undecanoic acid
C11H22O2
MFCD00002730
hendecanoic acid, undecylic acid, n-undecanoic acid, n-undecoic acid, n-undecylic acid, undecoic acid, undecanoate, 1-decanecarboxylic acid, undekansaeure, unii-138on3iiqg
CHEBI:32368
CCCCCCCCCCC(=O)O

Specifications

112-37-8
White to Yellow
>110°C
98.5% min. (GC)
C11H22O2
MFCD00002730
02, 358
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,cresols and 1-butanol
CCCCCCCCCCC(=O)O
186.29
CHEBI:32368
99%
Undecanoic acid, 99%
28.5°C
284.0°C
Authentic
Glass bottle
CH3(CH2)9CO2H
5 g
hendecanoic acid, undecylic acid, n-undecanoic acid, n-undecoic acid, n-undecylic acid, undecoic acid, undecanoate, 1-decanecarboxylic acid, undekansaeure, unii-138on3iiqg
ZDPHROOEEOARMN-UHFFFAOYSA-N
undecanoic acid
8180
186.29
Low Melting Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-964-2

RTECSNumber : YQ2275000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only