missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Undecanoyl Chloride 97.0+%, TCI America™

Supplier:  TCI America U002610ML

 View more versions of this product

Catalog No. U002610ML


Explore available promotions

Description

Description

Specifications

Specifications

Undecanoyl Chloride
Yellow
C11H21ClO
10 mL
n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f
CCCCCCCCCCC(=O)Cl
204.738
204.74
Liquid
17746-05-3
130°C
MFCD00000739
3265
JUKPJGZUFHCZQI-UHFFFAOYSA-N
undecanoyl chloride
87287
≥97.0% (GC,T)

Chemical Identifiers

17746-05-3
204.738
JUKPJGZUFHCZQI-UHFFFAOYSA-N
87287
CCCCCCCCCCC(=O)Cl
C11H21ClO
MFCD00000739
n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f
undecanoyl chloride
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions