Learn More
Thermo Scientific™ Uniguard™ Direct-Connection Guard Cartridge Holders
Eliminate the requirement for extra fittings with Thermo Scientific™ Uniguard Direct-Connection Guard Cartridge Holders.
$316.76 - $439.72
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | |||||
85000
|
Thermo Scientific™
85000 |
4.6 mm I.D. Guard Columns |
Each for $439.72
|
|
|||||
03-052-393
|
Thermo Scientific™
850WT |
Waters Columns Replacement Tips |
Each for $316.76
|
|
|||||
85100
|
Thermo Scientific™
85100 |
1 mm I.D. Guard Columns |
Each for $410.00
|
|
|||||
85200
|
Thermo Scientific™
85200 |
2.1 to 3 mm I.D. Guard Columns |
Each for $435.77
|
|
|||||
Description
Eliminate the requirement for extra fittings using Thermo Scientific™ Uniguard Direct-Connection Guard Cartridge Holders. They are reusable, stainless-steel guard cartridge holders that attach directly to the analytical column inlet.
- With PEEK 0.0625in. male outlet that fits many columns
- 0.0625in. female inlet tip can be used with various standard fittings
Ordering Information:
Other column dimensions are available. Please call your Customer Service Representative for more information.
Specifications
Cartridge Holder |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.