United Scientific™ Clamp Holder
Clamp Holder for rods up to 19 mm in diameter.
Supplier:  United Scientific™ RCLBH1
Catalog No. S13071
Explore available promotions
Description
Description
  • Made of cast aluminum body with steel thumb screws.
Specifications
Specifications
Rod Connector
Rods up to 19 mm in dia.
Single Adjustment
Cast Aluminum (Body), Steel (Thumb Screws)
Silver