United Scientific™ Secchi Disk
Use this Secchi Disk for water clarity measurements.
Supplier:  United Scientific™ SCDSK1
Catalog No. S12871
Explore available promotions
Description
Description
  • Good quality and inexpensive Secchi Disk for water clarity measurements.
  • Constructed of robust white plastic disk with permanent screen-printed black sectors.
  • Includes attached sinker and rot-proof polymer line on polymer foam bobbin.
  • Line length is approximately 72 feet (22 m) and includes a marking at each foot.
Specifications
Specifications
72 ft.
Secchi Disk
22 m
K+