missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
United Scientific™ Hand Lens
Glass Hand Lens mounted on a metal frame.
$14.41 - $14.41
Specifications
Magnification | 2.5x |
---|---|
Material | Glass |
Product Type | Lens, Hand |
Includes | Bakelite Handle |
Catalog Number | Mfr. No. | Diameter (English) Outer | Diameter (Metric) Outer | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) Outer | Diameter (Metric) Outer | Price | Quantity | |||||
36-104-5589
![]() |
United Scientific™
MFG075 |
2.95 in. | 7.5 cm |
Each for $14.41
|
|
|||||
Description
- Magnifier
- With bakelite handles
Specifications
2.5x | |
Glass | |
Lens, Hand | |
Bakelite Handle |