Beckman Coulter Universal Liquids Module (ULM) for LS 13 320 XR Particle Size Analyzer
The Universal Liquids Module (ULM) is a sampling module for use with the LS 13 320 XR Particle Size Analyzer. It measures the entire sample introduced to the instrument by re-circulating the sample, and requires relatively small volumes of sample and dispersing liquid (125 mL).
Supplier: Beckman Coulter B98105
Description
- Analyzes samples suspended in aqueous as well as non-aqueous dilutents for maximum flexibility
- Analytical size range: 10 nm - 2,000 μm (Optical bench dependant)
- Optional ULM Sonication Unit available
Specifications
Sampling Module | |
0.010 μm to 2,000 μm | |
LS 13 320 XR Particle Size Analyzer |
125 mL | |
Particle Size Analysis of Samples Suspended in Aqueous and Non-Aqueous Solutions |