missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Globe Scientific Universal Pipet Tips
Available as non-sterile
$72.94 - $73.61
Specifications
Sterility | Non-sterile |
---|---|
Autoclavable | Autoclavable |
Format | Bulk |
Tip Style | Universal |
For Use With (Equipment) | Universal Fit |
Catalog Number | Mfr. No. | Volume (Metric) | Color | Sterility | Format | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Color | Sterility | Format | Price | Quantity | ||
22-170-173
|
Globe Scientific
151150 |
1 to 200 μL | Natural | Non-sterile | Bulk |
Pack of 1000 Each for $73.61
Case of 10 Pack for $736.10
|
|
||
22-170-403
|
Globe Scientific
151146 |
100 to 1000 μL | Blue | Non-sterile | Bulk |
Pack of 1000 Each for $72.94
|
|
||
Description
- Made from prime virgin polypropylene
- Universal fit, used with many pipets
- Reference tips feature graduation marks for precise measurements
- Autoclavable (121°C at 15 minutes)
Specifications
Non-sterile | |
Bulk | |
Universal Fit | |
Polypropylene |
Autoclavable | |
Universal | |
Yes |