Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Secure vessels using these universal platform clamps for orbital shakers.
Supplier: Thermo Scientific™ 236028
Description
Secure vessels using Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers. With exceptional innovation, reliability and accuracy, these orbital shakers, platforms and accessories are the optimal choice for a busy lab.
Flask Clamp Starter Kit includes:
- (1) microplate clamp
- (2) 25mL flask clamps
- (2) 50mL flask clamps
- (2) 125mL flask clamps
- (4) 250/300mL flask clamps
- (4) 500mL flask clamps
- (2) 1L flask clamps
- (2) 2L flask clamps
- Replacement screws
Specifications
5 L | |
5L Erlenmeyer Flask Clamp | |
0 |
Flask Clamp | |
MaxQ HP 416, 420, 430, 435, 436, 480, 481 |