missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Universol- ES liquid scintillation cocktail, MP Biomedicals™

$563.04 - $1612.31

Specifications

Chemical Name or Material Universol- ES liquid scintillation cocktail
Density 0.9 to 1.0g/mL at 20°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN88248001
View Documents
MP Biomedicals Inc
0188248001
1 gal.
Each for $563.04
 
ICN88248005
View Documents
MP Biomedicals Inc
0188248005
5 gal.
Each for $1,612.31
 
Description

Description

Specifications

Specifications

Universol- ES liquid scintillation cocktail
0.9 to 1.0g/mL at 20°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'