Idex Micro Tight™ Nuts DFS Item
Fittings for very high pressure applications
Supplier:  Idex UPP416
Catalog No. 29-008-329
Explore available promotions
Description
Description
  • Ideal for UHPLC applications Quick component replacement with minimal down time
  • Max pressure of 15,000 psi (1035 bar)
  • Requires 4.0 in-lb (0.45 N-m) of torque
  • Natural PEEK
Specifications
Specifications
PEEK