Idex Plastic Tubing Cutter DFS Item
Spring-loaded guillotine cutters designed specifically for semi-rigid and rigid polymer tubing
Supplier:  Idex UPA327
 View more versions of this product
Catalog No. 05-701-84
Explore available promotions
Description
Description
Spring-loaded guillotine cutters designed specifically for semi-rigid and rigid polymer tubing
  • Built-in guide holes for both 1/16 and 1/8 in. OD tubing
  • Replacement blades included and sold separately
Flat, 90°, burr-free cut
Offers durability, reliability and ease
  • Able to cut through polymers such as PEEK™, FEP, PFA and Tefzel™
  • Specifications
    Specifications
    Tubing Cutter
    1/16, 1/8 in.
    Upchurch™ standard polymer tubing cutter
    Plastic
    1.59, 3.175 mm
    Black