missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 241020010

 View more versions of this product

Catalog No. AC241020010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

Urea hydrogen peroxide
75.0°C to 85.0°C
Glass bottle
CO(NH2)2·H2O2
16,379; 17,401
urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid
AQLJVWUFPCUVLO-UHFFFAOYSA-N
hydrogen peroxide;urea
1% sodium pyrophosphate
CHEBI:75178
≥33.5% (H2O2)
124-43-6,7722-88-5
White
CH6N2O3
1 kg
15,1784
Solubility in water: 500g/L (20°C). Other solubilities: alcohol and ether decomposes the product to h2o2, and urea, soluble in ethylene glycol
C(=O)(N)N.OO
94.07
31294
94.07
Tablets

Chemical Identifiers

124-43-6
94.07
urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid
CHEBI:75178
C(=O)(N)N.OO
CH6N2O3
AQLJVWUFPCUVLO-UHFFFAOYSA-N
31294
hydrogen peroxide;urea
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 204-701-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only