missing translation for 'onlineSavingsMsg'
Learn More

Uridine-5'-diphosphate disodium salt, 98%

CAS: 27821-45-0 | C9H12N2Na2O12P2 | 448.12 g/mol

Supplier:  Thermo Scientific Chemicals J6432903

 View more versions of this product

Catalog No. AAJ6432903


Only null left
Explore available promotions

Description

Description

Uridine-5'-diphosphate disodium salt is used in studies on nucleic acid (RNA) biosynthesis and cell signaling. UDP is a nucleotide that upon phosphorylation to UTP becomes a substrate for enzymes such as RNA polymerase(s) and GTPases. These enzymes are involved in a wide range of applications from the synthesis of RNA to the regulation of G-coupled Protein Receptors (GPCR) and cell signaling molecules such as Rho-signaling via Guanine Nucleotide Exchange Factors (GEF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Uridine-5′-diphosphate disodium salt is used in studies on nucleic acid (RNA) biosynthesis and cell signaling. UDP is a nucleotide that upon phosphorylation to UTP becomes a substrate for enzymes such as RNA polymerase(s) and GTPases. These enzymes are involved in a wide range of applications from the synthesis of RNA to the regulation of G-coupled Protein Receptors (GPCR) and cell signaling molecules such as Rho-signaling via Guanine Nucleotide Exchange Factors (GEF).

Solubility
Slightly soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
Specifications

Specifications

Uridine-5'-diphosphate disodium salt
White
MFCD00006524,MFCD00084679,MFCD04975448
8817400
5'-udp,2na
ZQKVPFKBNNAXCE-HVQNCJTOSA-L
disodium;(2R,3R,4S,5R)-2-(2,4-dioxopyrimidin-1-yl)-5-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolane-3,4-diolate
131676107
98%
27821-45-0
C9H12N2Na2O12P2
1 g
Moisture sensitive
Slightly soluble in water.
[Na+].[Na+].OC1C(O)[C@@H](O[C@@H]1COP(O)(=O)OP([O-])([O-])=O)N1C=CC(=O)NC1=O
448.12
448.13
Powder

Chemical Identifiers

27821-45-0
448.12
ZQKVPFKBNNAXCE-HVQNCJTOSA-L
131676107
C9H12N2Na2O12P2
MFCD00006524,MFCD00084679,MFCD04975448
5'-udp,2na
[Na+].[Na+].OC1C(O)[C@@H](O[C@@H]1COP(O)(=O)OP([O-])([O-])=O)N1C=CC(=O)NC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 248-678-9

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only