Uridine-5'-Diphosphoglucose Disodium Salt , MP Biomedicals™
Uridine-5'-diphosphoglucose Disodium Salt is a biosynthetic product produced by the reaction of UTP and glucose-1-phosphate catalyzed by uridyl transferase.
$147.74 - $147.74
Specifications
Quantity 100 mg
Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MP210120880
View Documents
MP Biomedicals Inc
0210120880
100 mg
Each for $147.74
Description
Description
  • Endogenous nucleotide sugar involved in glycosyltransferase reactions in metabolism
  • Shown to bind the P2Y14 receptor, an atypical P2Y receptor involved in the activation of dendritic cells and glial cells
  • Binds to and activates GPR17, inducing oligodendrocyte differentiation at a maximal concentration of 100μM
Specifications
Specifications
100 mg
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'