Ascensia Diabetes Care Urinalysis Reagent Test Strips
Single-use, plastic strips with multiple reagent test areas
Supplier:  Ascensia Diabetes Care 2806
 View more versions of this product
Catalog No. 36-099-4167
Explore available promotions
Description
Description
  • Ready to use and entirely disposable
  • Dip and read visually for results
  • Some strips can be read instrumentally on Clinitek™ 50, Clinitek 100, Clinitek 200, Clinitek 200+ and Clinitek 500 Urine Chemistry analyzers
Specifications
Specifications
Waived
Single-use
50 Pack
Glucose
Urinalysis