missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Urocanic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 228960250

Catalog No. AC228960250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Urocanic acid
0.5% max.
97.5% min. (HPLC)
C6H6N2O2
25g
10, 9691
Solubility in water: slightly soluble. Other solubilities: soluble in acetone,,insoluble in ethanol,diethyl ether
C1=C(NC=N1)C=CC(=O)O
138.13
CHEBI:30818
98%
104-98-3
Authentic
Glass bottle
MFCD00005203
25, 126
urocanic acid, 4-imidazoleacrylic acid, trans-urocanic acid, urocaninic acid, imidazoleacrylic acid, 5-imidazoleacrylic acid, urocanate, imidazole-4-acrylic acid, 2e-3-1h-imidazol-4-yl acrylic acid, 3-1h-imidazol-4-yl-2-propenoic acid
LOIYMIARKYCTBW-UPHRSURJSA-N
(Z)-3-(1H-imidazol-5-yl)prop-2-enoic acid
1549103
138.13
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-258-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only