Honeywell™ Uvex™ Astro™ Rx 3003 Safety Glasses
$214.68 - $214.68
Specifications
Lens Tint Clear
Frame Material Nylon
View More Specs
Products 1
Catalog Number Mfr. No. Frame Color Price Quantity  
Catalog Number Mfr. No. Frame Color Price Quantity  
18-079F
View Documents
Honeywell™
UVS2572
Blue
Pack of 10 Each for $214.68
Description
Description
The astro Rx 3003 spectacles feature the styling and fit of astrospec 3000™, plus a bevel groove for prescription lenses and unique lens carrier replacement system.Lightweight, impact-resistant nylon frames are available in black; blue; yellow and black; and red, white, and blue. Propionate browguard and sideshields provide protection against UV absorption, dust, and flying particles.
Order Info
See frame color guide for color selection.
Specifications
Specifications
Clear
Nylon
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'