Honeywell™ Uvex™ Bionic™ Face Shields
Designed for rugged jobs—tough yet lightweight and comfortable
Supplier:  Honeywell™ UVS8510
Catalog No. 29-635-3813
Explore available promotions
Description
Description
For workers exposed to falling or flying objects, impact, chemical splash, and airborne debris. Fit with most goggles and respirators.
  • Shell offers built-in chin protection and extended top-of-head coverage
  • Adjustable headgear tilts visor into 2784 possible positions
  • Ratcheting headgear features a locking mechanism for secure fit
  • Comfortable cell foam on back of headgear with breathable, removable, washable headband
  • 100% dielectric; no metal parts
Replacement Visors
  • Clear lens suits most indoor work applications
  • Shade 5.0 suits welding, brazing, and cutting
Compliance
Meets ANSI Z87.1+ and CSA Z94.3 standards.