Honeywell™ Uvex™ Classic™ Chemical-Splash Goggles
Integrated ventilation system
Supplier:  Honeywell™ UVS360
Catalog No. 17-253
Explore available promotions
Description
Description
  • Clear polycarbonate lens is treated with patented uvextreme™ anti-fog coating(XTR) to protect against fogging, scratching, static dust, and 99% of harmful ultraviolet rays up to 385nm
  • Large, soft body ensures excellent fit, even over most prescription spectacles
  • Indirect ventilation system allows air to flow freely through goggle body
  • Wide neoprene headstrap withstands acids, caustics, solvents, and alcohols
Compliance
Meets ANSI Z87.1-2010 and CSA Z94.3 standards.
Specifications
Specifications
UV extreme
Chemical Splash Protection Goggles
Polycarbonate
Clear
Wide neoprene headstrap frame
Product Suggestions
Product Suggestions