Honeywell™ Uvex™ Flex Seal™ Goggles
Exceptionally tight seal for protection from impacts and chemical splashes.
Supplier:  Honeywell™ UVS3430X
 View more versions of this product
Catalog No. 191300835
Explore available promotions
Description
Description
  • Contemporary
  • Low-profile
  • With wrap-around
  • Optically correct polycarbonate lens configuration increases visibility and protection
  • Uvextreme™ antifog coating and indirect ventilation minimize fogging
  • Temperature-resistant blue silicone frames fit over virtually all prescription eyewear
  • Pivoting clips permit goggle to be positioned on hardhat or on headband above the ears
Order Info
Available with neoprene band or fabric band.
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards.
Specifications
Specifications
Anti-fog
Shade 3
Blue
Neoprene headband
Polycarbonate
Uvextreme Anti-fog
Safety Goggles
Neoprene
Indirect
Silicone